2,5-Dimethyl-2-hexanol ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
B20352.06
B20352.14
B20352.06EA
41.2
EUR
B20352.06
B20352.14
2,5-Dimethyl-2-hexanol ≥98%
2,5-Dimethyl-2-hexanol
Purity: 98%
CAS No.: 3730-60-7
Molecular Formula: C8H18O
Formula Weight: 130.23
Flash Point: 52°(125°F)
Refractive Index: 1.4220
Boiling Point: 155-156°
Beilstein Reference: 1732761
UN No.: 1987
MDL No.: MFCD00021814
Formel:
C₈H₁₈O Molecular Weight: 130.23 g/mol Siedepunkt: 155…156 °C Dichte: 0.8203 g/cm³ (0 °C) Flammpunkt: 52 °C (125 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00021814 CAS-Nummer: 3730-60-7 EINECS: 223-085-8 UN: 1987 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...