Ethyl-2-chlornicotinat ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Ethyl 2-chloropyridine-3-carboxylate, Ethyl 2-Chloro-3-pyridinecarboxylate, Ethyl 2-chloronicotinate

B20359.09 B20359.18
B20359.09EA 55.6 EUR
B20359.09 B20359.18
Ethyl-2-chlornicotinat ≥98%
Ethyl-2-chlornicotinat

Purity: 98% CAS No.: 1452-94-4 Molecular Formula: C8H8ClNO2 Formula Weight: 185.61 MDL No.: MFCD00674129

Formel: C₈H₈ClNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00674129
CAS-Nummer: 1452-94-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR