1-Cyclohexylethanol ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
B20363.06
B20363.14
B20363.06EA
98.8
EUR
B20363.06
B20363.14
1-Cyclohexylethanol ≥98%
1-Cyclohexylethanol
Purity: 98%
CAS No.: 1193-81-3
Molecular Formula: C8H16O
Formula Weight: 128.22
Flash Point: 73°(163°F)
Refractive Index: 1.4650
Boiling Point: 188-190°
MDL No.: MFCD00001475
Formel:
C₈H₁₆O Siedepunkt: 188…190 °C Dichte: 0,92 Flammpunkt: 73 °C (163 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001475 CAS-Nummer: 1193-81-3 EINECS: 214-780-7 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...