2,4,5-Trifluorphenol ≥94%
Lieferant: Thermo Scientific
|
Gefahr
|
B20395.03
B20395.03EA
46
EUR
B20395.03
2,4,5-Trifluorphenol ≥94%
2,4,5-Trifluorphenol
Purity: 94%
CAS No.: 2268-16-8
Molecular Formula: C6H3F3O
Formula Weight: 148.09
Flash Point: 55°(131°F)
Odour: Phenol-like
UN No.: 2921
MDL No.: MFCD00061215
Formel:
C₆H₃F₃O Molecular Weight: 148,08 g/mol Siedepunkt: 155 °C (1013 hPa) Schmelzpunkt: 39…42 °C Flammpunkt: 55 °C (131 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00061215 CAS-Nummer: 2268-16-8 UN: 2921 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...