2-(2-Naphthyl)indol ≥98%

Lieferant: Thermo Fisher Scientific

Synonyme: 2-(Naphthalen-2-yl)-1H-indole

B20434.03 B20434.06
B20434.03EA 84.8 EUR
B20434.03 B20434.06
2-(2-Naphthyl)indol ≥98%
2-(2-Naphthyl)indole

Purity: 98% CAS No.: 23746-81-8 Molecular Formula: C18H13N Formula Weight: 243.31 MDL No.: MFCD00124144

Formel: C₁₈H₁₃N
Molecular Weight: 243.31 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00124144
CAS-Nummer: 23746-81-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR