2-(4-Pyridinyl)-4-thiazolcarbonsäure ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

B20457.03 B20457.06
B20457.03EA 37.1 EUR
B20457.03 B20457.06
2-(4-Pyridinyl)-4-thiazolcarbonsäure ≥97%
2-(4-Pyridinyl)-4-thiazolcarbonsäure

Purity: 97% CAS No.: 21278-86-4 Molecular Formula: C9H6N2O2S Formula Weight: 206.22 Melting Point: 300° dec. MDL No.: MFCD00171745

Formel: C₉H₆N₂O₂S
Molecular Weight: 206.22 g/mol
Schmelzpunkt: >300 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00171745
CAS-Nummer: 21278-86-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR