Methyl-5-brom-2-furoat ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

B20524.06 B20524.03 B20524.14
B20524.06EA 87.5 EUR
B20524.06 B20524.03 B20524.14
Methyl-5-brom-2-furoat ≥97%
Methyl-5-brom-2-furoat

Purity: 97% CAS No.: 2527-99-3 Molecular Formula: C6H5BrO3 Formula Weight: 205.01 Melting Point: 62-65° MDL No.: MFCD00092313

Formel: C₆H₅BrO₃
Schmelzpunkt: 62…65 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00092313
CAS-Nummer: 2527-99-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR