Methyl-5-brom-2-furoat ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
B20524.06
B20524.03
B20524.14
B20524.06EA
87.5
EUR
B20524.06
B20524.03
B20524.14
Methyl-5-brom-2-furoat ≥97%
Methyl-5-brom-2-furoat
Purity: 97%
CAS No.: 2527-99-3
Molecular Formula: C6H5BrO3
Formula Weight: 205.01
Melting Point: 62-65°
MDL No.: MFCD00092313
Formel:
C₆H₅BrO₃ Schmelzpunkt: 62…65 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00092313 CAS-Nummer: 2527-99-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...