(2,4,6-Trimethylphenyl)acetonitril ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
2,4,6-Trimethylbenzylcyanid, Mesitylacetonitril
B20534.06
B20534.06EA
79.9
EUR
B20534.06
(2,4,6-Trimethylphenyl)acetonitril ≥98%
(2,4,6-Trimethylphenyl)acetonitril
Purity: 98%
CAS No.: 34688-71-6
Molecular Formula: C11H13N
Formula Weight: 159.23
Melting Point: 77-80°
Boiling Point: 120°/1mm
UN No.: 3439
MDL No.: MFCD00013815
Formel:
C₁₁H₁₃N Molecular Weight: 159,23 g/mol Siedepunkt: 120 °C (1 mmHg) Schmelzpunkt: 77…80 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00013815 CAS-Nummer: 34688-71-6 EINECS: 252-151-9 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...