2,6-Dichlorisonicotinsäure ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2,6-Dichloropyridine-4-carboxylic acid
B20541.06
B20541.06EA
146
EUR
B20541.06
2,6-Dichlorisonicotinsäure ≥98%
2,6-Dichlorisonicotinsäure
Purity: 98%
CAS No.: 5398-44-7
Molecular Formula: C6H3Cl2NO2
Formula Weight: 192.00
Melting Point: 208° dec.
MDL No.: MFCD00234147
Formel:
C₆H₃Cl₂NO₂ Molecular Weight: 192 g/mol Schmelzpunkt: 208 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00234147 CAS-Nummer: 5398-44-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...