4-Methyl-1H-imidazole-5-carbaldehyde 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-Methyl-5-imidazolecarboxaldehyde

B20608.06
B20608.06EA 104 EUR
B20608.06
4-Methyl-1H-imidazole-5-carbaldehyde 98%
4-Methyl-1H-imidazole-5-carbaldehyde

Purity: 99% CAS No.: 68282-53-1 Molecular Formula: C5H6N2O Formula Weight: 110.12 Melting Point: 166-170° MDL No.: MFCD00173728

Formel: C₅H₆N₂O
Molecular Weight: 110,12 g/mol
Schmelzpunkt: 166…170 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00173728
CAS-Nummer: 68282-53-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR