3-Ethyl-2-methyl-1-pentene 99%

Lieferant: Thermo Fisher Scientific
Gefahr

B20624.06
B20624.06EA 82.1 EUR
B20624.06
3-Ethyl-2-methyl-1-pentene 99%
3-Ethyl-2-methyl-1-pentene

Purity: 99% CAS No.: 19780-66-6 Molecular Formula: C8H16 Formula Weight: 112.22 Refractive Index: 1.4124 UN No.: UN3295 MDL No.: MFCD00048664

Formel: C₈H₁₆
Dichte: 0,72
Lagertemperatur: Raumtemperatur
MDL: MFCD00048664
CAS-Nummer: 19780-66-6
ADR: 3,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR