4-Hydroxy-3-methoxybenzylamin Hydrochlorid 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
B20637.06
B20637.14
B20637.06EA
73.5
EUR
B20637.06
B20637.14
4-Hydroxy-3-methoxybenzylamin Hydrochlorid 98%
4-Hydroxy-3-methoxybenzylamin Hydrochlorid
Purity: 98%
CAS No.: 7149-10-2
Molecular Formula: C8H11NO2.HCl
Formula Weight: 189.64
Melting Point: ca 220° dec.
Beilstein Reference: 3915418
Formel:
C₈H₁₂ClNO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012864 CAS-Nummer: 7149-10-2 EINECS: 230-468-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...