1,3-Diphenyl-1-butanon 95%

Lieferant: Thermo Scientific

B20654.03
B20654.03EA 57.2 EUR
B20654.03
1,3-Diphenyl-1-butanon 95%
1,3-Diphenyl-1-butanon

Purity: 95% CAS No.: 1533-20-6 Molecular Formula: C16H16O Formula Weight: 224.30 Melting Point: 68-72° MDL No.: MFCD00026345

Formel: C₁₆H₁₆O
Molecular Weight: 224,30 g/mol
Schmelzpunkt: 68…72 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00026345
CAS-Nummer: 1533-20-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR