Methyl-3,5-dibrombenzoat ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Methyl 3,5-dibromobenzoate
B20668.06
B20668.06EA
83
EUR
B20668.06
Methyl-3,5-dibrombenzoat ≥98%
Methyl-3,5-dibrombenzoat
Purity: 98%
CAS No.: 51329-15-8
Molecular Formula: C8H6Br2O2
Formula Weight: 293.94
Melting Point: 61-64°
Beilstein Reference: 2255855
Formel:
C₈H₆Br₂O₂ Molecular Weight: 293,94 g/mol Schmelzpunkt: 61…64 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00082648 CAS-Nummer: 51329-15-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...