1-Pentansulfonsäure Natriumsalz, (max. 1% H₂O) 99% (Trockengewicht)
Lieferant: Thermo Fisher Scientific
Synonyme:
Natriumpentylsulfonat
B20699.06
B20699.14
B20699.22
B20699.06EA
44.5
EUR
B20699.06
B20699.14
B20699.22
1-Pentansulfonsäure Natriumsalz, (max. 1% H₂O) 99% (Trockengewicht)
1-Pentanesulphonic acid sodium salt
Purity: 99%
CAS No.: 22767-49-3
Molecular Formula: CH3(CH2)4SO3Na
Formula Weight: 174.19
Melting Point:
Formel:
C₅H₁₁O₃SNa Molecular Weight: 174.2 g/mol Schmelzpunkt: >300 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007541 CAS-Nummer: 22767-49-3 EINECS: 245-208-4 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...