8-Hexadecanol ≥97%

Lieferant: Thermo Scientific
Achtung

B20734.22
B20734.22EA 0 EUR
B20734.22
8-Hexadecanol ≥97%
8-Hexadecanol

Purity: 97% CAS No.: 19781-83-0 Molecular Formula: CH3(CH2)7CH(OH)(CH2)6CH3 Formula Weight: 242.45 MDL No.: MFCD00046733

Formel: C₁₆H₃₄O
Molecular Weight: 242,45 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00046733
CAS-Nummer: 19781-83-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR