5-(4-Pyridyl)-1H-tetrazol ≥98%
Lieferant: Thermo Scientific
Synonyme:
4-(1H-Tetrazol-5-yl)pyridine
B20741.03
B20741.03EA
78.5
EUR
B20741.03
5-(4-Pyridyl)-1H-tetrazol ≥98%
5-(4-Pyridyl)-1H-tetrazole
Purity: 98%
CAS No.: 14389-12-9
Molecular Formula: C6H5N5
Formula Weight: 147.14
MDL No.: MFCD00068115
Formel:
C₆H₅N₅ Molecular Weight: 147,14 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00068115 CAS-Nummer: 14389-12-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...