2,4-Chinolindiol ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

B20747.06 B20747.14
B20747.06EA 23.4 EUR
B20747.06 B20747.14
2,4-Chinolindiol ≥97%
2,4-Chinolindiol

Purity: 97% CAS No.: 86-95-3 Molecular Formula: C9H7NO2 Formula Weight: 161.16 Melting Point: 300° Solubility: Insoluble in water Beilstein Reference: 129767 MDL No.: MFCD00006744

Formel: C₉H₇NO₂
Siedepunkt: 375 °C (1013 hPa)
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006744
CAS-Nummer: 86-95-3
EINECS: 201-711-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR