2-Ethylcyclohexanon ≥99%

Lieferant: Thermo Fisher Scientific

B20748.06 B20748.14
B20748.06EA 76.4 EUR
B20748.06 B20748.14
2-Ethylcyclohexanon ≥99%
2-Ethylcyclohexanone

Purity: 99% CAS No.: 4423-94-3 Molecular Formula: C8H14O Formula Weight: 126.20 MDL No.: MFCD00066306

Formel: C₈H₁₄O
Lagertemperatur: Raumtemperatur
MDL: MFCD00066306
CAS-Nummer: 4423-94-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR