Methyl-4,5-dibromo-3-hydroxythiophene-2-carboxylate 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: Methyl 4,5-dibromo-3-hydroxythiophene-2-carboxylate

B20780.06
B20780.06EA 88 EUR
B20780.06
Methyl-4,5-dibromo-3-hydroxythiophene-2-carboxylate 97%
Methyl-4,5-dibromo-3-hydroxythiophene-2-carboxylate

Purity: 97% CAS No.: 96232-71-2 Molecular Formula: C6H4Br2O3S Formula Weight: 316.89 Melting Point: 125-128° MDL No.: MFCD00052082

Formel: C₆H₄Br₂O₃S
Molecular Weight: 316,89 g/mol
Schmelzpunkt: 125…128 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00052082
CAS-Nummer: 96232-71-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR