1,1-Cyclohexandiessigsäure ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

B20816.09 B20816.18
B20816.09EA 48.2 EUR
B20816.09 B20816.18
1,1-Cyclohexandiessigsäure ≥98%
1,1-Cyclohexandiessigsäure

Purity: 98% CAS No.: 4355-11-7 Molecular Formula: C10H16O4 Formula Weight: 200.23 Melting Point: 178-182° MDL No.: MFCD00001519

Formel: C₁₀H₁₆O₄
Schmelzpunkt: 178…182 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00001519
CAS-Nummer: 4355-11-7
EINECS: 224-427-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR