5-Fluorisatin ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
B20831.03
B20831.06
B20831.14
B20831.03EA
22.6
EUR
B20831.03
B20831.06
B20831.14
5-Fluorisatin ≥98%
5-Fluorisatin
Purity: 98%
CAS No.: 443-69-6
Molecular Formula: C8H4FNO2
Formula Weight: 165.12
Melting Point: 224-227°
Beilstein Reference: 131241
Formel:
C₈H₄FNO₂ Schmelzpunkt: 224…227 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00022795 CAS-Nummer: 443-69-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...