2-(Trifluormethoxy)benzaldehyd 96%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
B20840.06
B20840.14
B20840.22
B20840.06EA
49.7
EUR
B20840.06
B20840.14
B20840.22
2-(Trifluormethoxy)benzaldehyd 96%
2-(Trifluormethoxy)benzaldehyd
Purity: 96%
CAS No.: 94651-33-9
Molecular Formula: C8H5F3O2
Formula Weight: 190.12
Flash Point: 67°(152°F)
Refractive Index: 1.4550
Boiling Point: 77°/20mm
Beilstein Reference: 6137162
MDL No.: MFCD00042405
Formel:
C₈H₅F₃O₂ Siedepunkt: 77 °C (20 mmHg) Dichte: 1,330 Flammpunkt: 67 °C (152 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00042405 CAS-Nummer: 94651-33-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...