3-Ethyl-3-hexanol ≥98%

Lieferant: Thermo Scientific
Achtung

B20859.06
B20859.06EA 115 EUR
B20859.06
3-Ethyl-3-hexanol ≥98%
3-Ethyl-3-hexanol

Purity: 98% CAS No.: 597-76-2 Molecular Formula: C8H18O Formula Weight: 130.23 MDL No.: MFCD00039616

Formel: C₈H₁₈O
Molecular Weight: 130,23 g/mol
Dichte: 0,83
Lagertemperatur: Raumtemperatur
MDL: MFCD00039616
CAS-Nummer: 597-76-2
EINECS: 209-908-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR