3-Methylbenzylalkohol ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3-(Hydroxymethyl)toluol, (3-Methylphenyl)methanol
B20875.14
B20875.14EA
62.5
EUR
B20875.14
3-Methylbenzylalkohol ≥98%
3-Methylbenzylalkohol
Purity: 98%
CAS No.: 587-03-1
Molecular Formula: C8H10O
Formula Weight: 122.17
Flash Point: 105°(221°F)
Refractive Index: 1.5340
Boiling Point: 216-217°
Beilstein Reference: 2324516
MDL No.: MFCD00004646
Formel:
C₈H₁₀O Molecular Weight: 122,17 g/mol Siedepunkt: 216…217 °C Schmelzpunkt: < –20 °C Dichte: 1,013 Flammpunkt: 105 °C (221 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004646 CAS-Nummer: 587-03-1 EINECS: 209-595-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...