5-(2,3-Dichlorphenyl)-1H-tetrazol ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
B20921.06
B20921.06EA
243
EUR
B20921.06
5-(2,3-Dichlorphenyl)-1H-tetrazol ≥97%
5-(2,3-Dichlorphenyl)-1H-tetrazol
Purity: 97%
CAS No.: 175205-12-6
Molecular Formula: C7H4Cl2N4
Formula Weight: 215.04
Melting Point: 180-182°
MDL No.: MFCD00068096
Formel:
C₇H₄Cl₂N₄ Molecular Weight: 215,04 g/mol Schmelzpunkt: 180…182 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00068096 CAS-Nummer: 175205-12-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...