3,4-Dimethylbenzophenon ≥99%

Lieferant: Thermo Fisher Scientific

Synonyme: (3,4-Dimethylphenyl)(phenyl)methanone

B20965.14 B20965.22
B20965.14EA 39.9 EUR
B20965.14 B20965.22
3,4-Dimethylbenzophenon ≥99%
3,4-Dimethylbenzophenon

Purity: 99% CAS No.: 2571-39-3 Molecular Formula: C15H14O Formula Weight: 210.28 Flash Point: 110°(230°F) Melting Point: 44-47° Boiling Point: 335° Beilstein Reference: 1952043 MDL No.: MFCD00008525

Formel: C₁₅H₁₄O
Siedepunkt: 335 °C
Schmelzpunkt: 44…47 °C
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008525
CAS-Nummer: 2571-39-3
EINECS: 219-916-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR