Biphenyl-4-sulfonylchlorid ≥97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
B21031.06
B21031.14
B21031.22
B21031.06EA
56.5
EUR
B21031.06
B21031.14
B21031.22
Biphenyl-4-sulfonylchlorid ≥97%
Biphenyl-4-sulfonylchlorid
Purity: 97%
CAS No.: 1623-93-4
Molecular Formula: C12H9ClO2S
Formula Weight: 252.72
Melting Point: 112-117°
Boiling Point: 200-205°/6mm
UN No.: 3261
MDL No.: MFCD00078658
Formel:
C₁₂H₉ClO₂S Siedepunkt: 200…205 °C (6 mmHg) Schmelzpunkt: 112…117 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00078658 CAS-Nummer: 1623-93-4 EINECS: 216-609-1 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...