2-Phenyl-1H-indole-3-carbaldehyde 97%

Lieferant: Thermo Scientific

Synonyme: 3-Formyl-2-phenylindole, 2-Phenylindole-3-carboxaldehyde

B21041.06
B21041.06EA 99.8 EUR
B21041.06
2-Phenyl-1H-indole-3-carbaldehyde 97%
2-Phenyl-1H-indole-3-carbaldehyde

Purity: 97% CAS No.: 25365-71-3 Molecular Formula: C15H11NO Formula Weight: 221.26 Melting Point: 249-253° MDL No.: MFCD00435481

Formel: C₁₅H₁₁NO
Molecular Weight: 221,26 g/mol
Schmelzpunkt: 249…253 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00435481
CAS-Nummer: 25365-71-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR