3-Methyl-1-benzofuran-2-carbonsäure 97%
Lieferant: Thermo Fisher Scientific
B21044.03
B21044.06
B21044.03EA
40.8
EUR
B21044.03
B21044.06
3-Methyl-1-benzofuran-2-carbonsäure 97%
3-Methyl-1-benzofuran-2-carbonsäure
Purity: 97%
CAS No.: 24673-56-1
Molecular Formula: C10H8O3
Formula Weight: 176.17
Melting Point: 192-197°
Beilstein Reference: 132104
MDL No.: MFCD00060511
Formel:
C₁₀H₈O₃ Schmelzpunkt: 192…197 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00060511 CAS-Nummer: 24673-56-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...