1,3-Propandiyl-bis(4-methylbenzolsulfonat) 98%

Lieferant: Thermo Scientific

Synonyme: Propane-1,3-diyl bis(4-methylbenzenesulphonate), Trimethylene Glycol Ditosylate, Trimethylene Glycol Bis(p-toluenesulphonate), 1,3-Bis(p-toluenesulphonyloxy)propane, 1,3-Propanediol Ditosylate

B21084.14
B21084.14EA 49.2 EUR
B21084.14
1,3-Propandiyl-bis(4-methylbenzolsulfonat) 98%
1,3-Bis(tosyloxy)propane

Purity: 98% CAS No.: 5469-66-9 Molecular Formula: C17H20O6S2 Formula Weight: 384.47 Melting Point: 90-92° MDL No.: MFCD00013392

Formel: C₁₇H₂₀O₆S₂
Molecular Weight: 384,46 g/mol
Schmelzpunkt: 90…92 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00013392
CAS-Nummer: 5469-66-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR