Phthalimidomethyl)-phosphonsäurediethylester 97%
Lieferant: Thermo Fisher Scientific
Synonyme:
Diethyl ((1,3-dioxoisoindolin-2-yl)methyl)phosphonate, (Phthalimidomethyl)phosphonic Acid Diethyl Ester
B21096.06
B21096.14
B21096.06EA
59.1
EUR
B21096.06
B21096.14
Phthalimidomethyl)-phosphonsäurediethylester 97%
Diethyl (phthalimidomethyl)phosphonate
Purity: 97%
CAS No.: 33512-26-4
Molecular Formula: C13H16NO5P
Formula Weight: 297.25
Melting Point: 60-64°
MDL No.: MFCD00134313
Formel:
C₁₃H₁₆NO₅P Schmelzpunkt: 60…64 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00134313 CAS-Nummer: 33512-26-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...