1,2-Bis(diphenylphosphino)benzol ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

B21106.01 B21106.03
B21106.01EA 39.5 EUR
B21106.01 B21106.03
1,2-Bis(diphenylphosphino)benzol ≥98%
1,2-Bis(diphenylphosphino)benzol

Purity: 98% CAS No.: 13991-08-7 Molecular Formula: C26H22P2 Formula Weight: 446.47 Melting Point: 185-189° MDL No.: MFCD00014081

Formel: C₃₀H₂₄P₂
Schmelzpunkt: 185…189 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00014081
CAS-Nummer: 13991-08-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR