Bis(dimethylamino)dimethylsilan 97%
Lieferant: Thermo Scientific
|
Gefahr
|
B21119.22
B21119.22EA
148
EUR
B21119.22
Bis(dimethylamino)dimethylsilan 97%
Bis(dimethylamino)dimethylsilan
Purity: 97%
CAS No.: 3768-58-9
Molecular Formula: [(CH3)2N]2Si(CH3)2
Formula Weight: 146.31
Flash Point: -7°(19°F)
Refractive Index: 1.4170
Melting Point: -98°
Boiling Point: 128-129°
Beilstein Reference: 1736095
UN No.: 2924
MDL No.: MFCD00008296
Formel:
C₆H₁₈N₂Si Molecular Weight: 146,31 g/mol Siedepunkt: 128…129 °C Schmelzpunkt: –98 °C Dichte: 0,807 Flammpunkt: –7 °C (19 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008296 CAS-Nummer: 3768-58-9 EINECS: 223-200-1 UN: 2924 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...