IPTG (Isopropyl-β-D-thiogalactopyranosid) ≥99% dioxanfrei

Lieferant: Thermo Fisher Scientific

Synonyme: IPTG

B21149.03 B21149.06
B21149.03EA 90.2 EUR
B21149.03 B21149.06
IPTG (Isopropyl-β-D-thiogalactopyranosid) ≥99% dioxanfrei
IPTG (Isopropyl-β-D-thiogalactopyranosid)

Purity: 99% CAS No.: 367-93-1 Molecular Formula: C9H18O5S Formula Weight: 238.30 Storage Temperature: -30°C to -10°C Melting Point: 109-115° Beilstein Reference: 4631 Optical Rotation: -32° (c=1 in water) MDL No.: MFCD00063273

Formel: C₉H₁₈O₅S
Molecular Weight: 238.31 g/mol
Siedepunkt: 402
Schmelzpunkt: 109…115 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00063273
CAS-Nummer: 367-93-1
EINECS: 206-703-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR