D-Threonin ≥99%
Lieferant: Thermo Fisher Scientific
B21177.06
B21177.14
B21177.22
B21177.06EA
40.1
EUR
B21177.06
B21177.14
B21177.22
D-Threonin ≥99%
D-Threonin
Purity: 99%
CAS No.: 632-20-2
Molecular Formula: C4H9NO3
Formula Weight: 119.12
Melting Point: ca 274° dec.
Beilstein Reference: 1721643
Formel:
C₄H₉NO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064269 CAS-Nummer: 632-20-2 EINECS: 211-171-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...