2-Octinsäuremethylester ≥98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: Methyl-2-octinoat

B21242.22
B21242.22EA 113 EUR
B21242.22
2-Octinsäuremethylester ≥98%
2-Octynoic acid methyl ester

Purity: 98% CAS No.: 111-12-6 Molecular Formula: CH3(CH2)4C≡CCO2CH3 Formula Weight: 154.20 Flash Point: 88°(192°F) Refractive Index: 1.4460 Boiling Point: 216-218° Beilstein Reference: 1756887 MDL No.: MFCD00009530

Formel: C₉H₁₄O₂
Molecular Weight: 154,21 g/mol
Siedepunkt: 216…218 °C
Dichte: 0,920
Flammpunkt: 88 °C (192 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00009530
CAS-Nummer: 111-12-6
EINECS: 203-836-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR