Diphenylmethan ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

B21256.0E B21256.22 B21256.36
B21256.0EEA 215 EUR
B21256.0E B21256.22 B21256.36
Diphenylmethan ≥99%
Diphenylmethan

Purity: 99+% CAS No.: 101-81-5 Molecular Formula: (C6H5)2CH2 Formula Weight: 168.24 Flash Point: 130°(266°F) Refractive Index: 1.5770 Melting Point: 22-26° Boiling Point: 262-264° Beilstein Reference: 1904982 Merck Reference: 14,3328 UN No.: 3077 MDL No.: MFCD00004781

Formel: C₁₃H₁₂
Siedepunkt: 262…264 °C
Schmelzpunkt: 22…26 °C
Dichte: 1,010
Flammpunkt: 130 °C (266 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00004781
CAS-Nummer: 101-81-5
EINECS: 202-978-6
UN: 3077
ADR: 9,III
Merck Index: 14,3328

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR