Serotonin Hydrochlorid ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 98%
CAS No.: 153-98-0
Molecular Formula: C10H12N2O.HCl
Formula Weight: 212.68
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Sensitivity: Light Sensitive and Hygroscopic
Appearance: White to pale cream
Melting Point: 167-172°
Beilstein Reference: 3570963
Merck Reference: 14,8463
UN No.: 2811
MDL No.: MFCD00012686
Formel:
C₁₀H₁₃ClN₂O Schmelzpunkt: 167…172 °C |
MDL:
MFCD00012686 CAS-Nummer: 153-98-0 EINECS: 244-464-4 UN: 2811 ADR: 6.1,III Merck Index: 14,8463 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...