Isochinolin 97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 2-Azanaphthalin

B21279.0E B21279.22 B21279.36
B21279.0EEA 969 EUR
B21279.0E B21279.22 B21279.36
Isochinolin 97%
Isochinolin

Purity: 97+% CAS No.: 119-65-3 Molecular Formula: C9H7N Formula Weight: 129.16 Flash Point: 107°(225°F) Refractive Index: 1.6245 Odour: Pungent pH: 7.5 Melting Point: 24-27° Boiling Point: 241-243° Beilstein Reference: 107549 Merck Reference: 14,5222 UN No.: 2811 MDL No.: MFCD00006898

Formel: C₉H₇N
Molecular Weight: 129.16 g/mol
Siedepunkt: 241…243 °C
Schmelzpunkt: 24…27 °C
Dichte: 1.090
Flammpunkt: 107 °C (225 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00006898
CAS-Nummer: 119-65-3
EINECS: 204-341-8
UN: 2811
ADR: 6.1 II
Merck Index: 13,05242

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR