6-Thioguanin 98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 6-Mercaptoguanine, 2-Amino-6-purinethiol, 2-Amino-3,7-dihydro-6H-purine-6-thione, 2-Amino-6-mercaptopurine

B21280.03
B21280.03EA 73.7 EUR
B21280.03
6-Thioguanin 98%
6-Thioguanin

Purity: 98% CAS No.: 154-42-7 Molecular Formula: C5H5N5S Formula Weight: 167.19 Odour: Odourless Melting Point: 360° Beilstein Reference: 157765 UN No.: UN2811 MDL No.: MFCD00233553

Formel: C₅H₅N₅S
Schmelzpunkt: >360 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00233553
CAS-Nummer: 154-42-7
EINECS: 205-827-2
ADR: 6.1,III
Merck Index: 14,9337

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR