tert-Butylchlordimethylsilan 50% (w/w) in Toluol

Lieferant: Thermo Scientific
Gefahr

Synonyme: tert-Butyl-chlordimethylsilan, Chlor-tert-butyldimethylsilan, tert-Butyldimethylchlorsilan, tert-Butyl(dimethyl)silylchlorid

B21286.18
B21286.18EA 121 EUR
B21286.18
tert-Butylchlordimethylsilan 50% (w/w) in Toluol
tert-Butylchlordimethylsilan

CAS No.: 18162-48-6 Molecular Formula: C6H15ClSi Formula Weight: 150.73 Flash Point: 7°(45°F) Odour: Characteristic UN No.: 2924 MDL No.: MFCD00000501

Formel: C₆H₁₅ClSi
Molecular Weight: 150,72 g/mol
Dichte: 0,87
Flammpunkt: 7 °C (45 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00000501
CAS-Nummer: 18162-48-6
EINECS: 242-042-4
UN: 2924
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR