N-(Hydroxymethyl)phthalimid ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: N-(Hydroxymethyl)phthalsäureimid, N-(Hydroxymethyl)-1,2-benzoldicarbonsäureimid

B21292.14 B21292.22 B21292.36
B21292.14EA 32.2 EUR
B21292.14 B21292.22 B21292.36
N-(Hydroxymethyl)phthalimid ≥97%
N-(Hydroxymethyl)phthalimid

Purity: 97% CAS No.: 118-29-6 Molecular Formula: C9H7NO3 Formula Weight: 177.16 Melting Point: 140-147° Beilstein Reference: 140946

Formel: C₉H₇NO₃
Siedepunkt: 415 °C (1013 hPa)
Schmelzpunkt: 140…147 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005899
CAS-Nummer: 118-29-6
EINECS: 204-241-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR