CAPSO (3-(Cyclohexylamino)-2-hydroxypropansulfonsäure) ≥98%

Lieferant: Thermo Fisher Scientific

Synonyme: 3-(Cyclohexylamino)-2-hydroxy-1-propanesulphonic acid

B21305.14 B21305.22 B21305.36
B21305.14EA 34.8 EUR
B21305.14 B21305.22 B21305.36
CAPSO (3-(Cyclohexylamino)-2-hydroxypropansulfonsäure) ≥98%
CAPSO (3-(Cyclohexylamino)-2-hydroxypropansulfonsäure)

Purity: 98% CAS No.: 73463-39-5 Molecular Formula: C9H19NO4S Formula Weight: 237.32 Melting Point: 270-274° MDL No.: MFCD00041778

Formel: C₉H₁₉NO₄S
Molecular Weight: 237.32 g/mol
Schmelzpunkt: 270…274 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00041778
CAS-Nummer: 73463-39-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR