1,2-Dichlorbutan ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
B21307.03
B21307.06
B21307.03EA
34.4
EUR
B21307.03
B21307.06
1,2-Dichlorbutan ≥97%
1,2-Dichlorobutane
Purity: 97+%
CAS No.: 616-21-7
Molecular Formula: C4H8Cl2
Formula Weight: 127.01
Flash Point: 30°(86°F)
Refractive Index: 1.4450
Boiling Point: 124°
Beilstein Reference: 1731376
UN No.: 1993
MDL No.: MFCD00013676
Formel:
ClH₂CCHClCH₂CH₃ Siedepunkt: 124 °C Dichte: 1,112 Flammpunkt: 30 °C (86 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00013676 CAS-Nummer: 616-21-7 EINECS: 210-469-5 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...