3,5-Difluorbenzonitril ≥99%

Lieferant: Thermo Fisher Scientific
Gefahr

B21347.06 B21347.14
B21347.06EA 38.9 EUR
B21347.06 B21347.14
3,5-Difluorbenzonitril ≥99%
3,5-Difluorbenzonitril

Purity: 99% CAS No.: 64248-63-1 Molecular Formula: C7H3F2N Formula Weight: 139.11 Melting Point: 83-88° Boiling Point: 160° UN No.: 3439 MDL No.: MFCD00010311

Formel: C₇H₃F₂N
Siedepunkt: 160 °C
Schmelzpunkt: 83…88 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00010311
CAS-Nummer: 64248-63-1
EINECS: 264-752-3
UN: 3439
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR