5-Methoxy-2-methylindol ≥99%

Lieferant: Thermo Fisher Scientific

B21348.03 B21348.06
B21348.03EA 39.5 EUR
B21348.03 B21348.06
5-Methoxy-2-methylindol ≥99%

Purity: 99+% CAS No.: 1076-74-0 Molecular Formula: C10H11NO Formula Weight: 161.20 Storage Temperature: 2°C to 8°C Melting Point: 85-88° MDL No.: MFCD00005620

Formel: C₁₀H₁₁NO
Siedepunkt: 145 °C (1,5 mmHg)
Schmelzpunkt: 85…88 °C
MDL: MFCD00005620
CAS-Nummer: 1076-74-0
EINECS: 214-066-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR