Kresolrot Natriumsalz
Lieferant: Thermo Fisher Scientific
B21361.14
B21361.22
B21361.14EA
62.3
EUR
B21361.14
B21361.22
Kresolrot Natriumsalz
Kresolrot Natriumsalz
CAS No.: 62625-29-0
Molecular Formula: C21H17NaO5S
Formula Weight: 404.42
Melting Point: ca 250° dec.
MDL No.: MFCD00001618
Formel:
C₂₁H₁₇NaO₅S Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001618 CAS-Nummer: 62625-29-0 EINECS: 263-654-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...