N-(2-Aminoethyl)morpholin ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
B21365.0B
B21365.18
B21365.30
B21365.0BEA
1372
EUR
B21365.0B
B21365.18
B21365.30
N-(2-Aminoethyl)morpholin ≥98%
N-(2-Aminoethyl)morpholin
Purity: 98+%
CAS No.: 2038-03-1
Molecular Formula: C6H14N2O
Formula Weight: 130.19
Flash Point: 86°(186°F)
Refractive Index: 1.4760
Odour: Amine-like
pH: 12
Melting Point: 23-25°
Boiling Point: 200-203°
Beilstein Reference: 104378
UN No.: 2923
MDL No.: MFCD00006182
Formel:
C₆H₁₄N₂O Siedepunkt: 200…203 °C Schmelzpunkt: 23…25 °C Dichte: 0,999 Flammpunkt: 86 °C (186 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006182 CAS-Nummer: 2038-03-1 EINECS: 218-011-6 UN: 2923 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...